siRNA Details
| Gene Symbol: Acsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108502.2 | NP_001101972.2 | 14 | 1785 | ||
| XM_006230127.2 | XP_006230189.1 | 15 | 1939 | |||
| XM_006230128.2 | XP_006230190.1 | 14 | 1855 | |||
| XM_006230129.2 | XP_006230191.1 | 14 | 1702 | |||
| XM_008759771.1 | XP_008757993.1 | 15 | 1855 | |||
| XM_008759772.1 | XP_008757994.1 | 14 | 1831 | |||
| XM_008759773.1 | XP_008757995.1 | 15 | 1935 | |||
| GenBank mRNA | BC168985.1 | - | 14 | 1785 | 7323876 | |
| FQ218416.1 | - | 10 | 978 | |||

Acsm1
Genomic Map