siRNA Details
| Gene Symbol: PEX11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184795.1 | NP_001171724.1 | 4 | 458 | ||
| NM_003846.2 | NP_003837.1 | 4 | 652 | |||
| NR_073491.1 | 4 | 648 | ||||
| NR_073492.1 | 4 | 642 | ||||
| NR_073493.2 | 4 | 864 | ||||
| GenBank mRNA | AB018080.1 | - | 4 | 434 | ||
| AF093670.1 | - | 4 | 432 | |||
| AK023991.1 | - | 4 | 416 | |||
| AK094173.1 | - | 1 | 1252 | |||
| AK301983.1 | - | 4 | 418 | |||
| BC011963.1 | - | 4 | 432 | 3866690 | ||
| BG423552.1 | - | 392 | ||||
| CB155824.1 | - | 4 | 458 | |||
| CR542047.1 | - | 4 | 416 | |||
| HY108328.1 | - | 4 | 435 | |||

PEX11B
Genomic Map