siRNA Details
| Gene Symbol: Prrt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276470.1 | NP_001263399.1 | 2 | 540 | ||
| XM_006230308.2 | XP_006230370.1 | 2 | 619 | |||
| GenBank mRNA | FQ213699.1 | - | 2 | 486 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prrt2| Gene Symbol: Prrt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276470.1 | NP_001263399.1 | 2 | 540 | ||
| XM_006230308.2 | XP_006230370.1 | 2 | 619 | |||
| GenBank mRNA | FQ213699.1 | - | 2 | 486 | ||