siRNA Details
| Gene Symbol: Armc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009455.1 | NP_001009455.1 | 5 | 2166 | ||
| GenBank mRNA | BC087086.1 | - | 5 | 2166 | 7189809 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Armc5| Gene Symbol: Armc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009455.1 | NP_001009455.1 | 5 | 2166 | ||
| GenBank mRNA | BC087086.1 | - | 5 | 2166 | 7189809 | |