siRNA Details
| Gene Symbol: Tial1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013193.1 | NP_001013211.1 | 9 | 952 | ||
| XM_006230310.2 | XP_006230372.1 | 9 | 1158 | |||
| XM_006230312.2 | XP_006230374.1 | 9 | 1209 | |||
| XM_006230313.2 | XP_006230375.1 | 9 | 1209 | |||
| XM_008759902.1 | XP_008758124.1 | 9 | 977 | |||
| GenBank mRNA | BC091409.1 | - | 9 | 952 | 7325481 | |

Tial1
Genomic Map