siRNA Details
| Gene Symbol: Plekha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079894.1 | NP_001073363.1 | 5 | 428 | ||
| XM_006230409.2 | XP_006230471.1 | 5 | 476 | |||
| XM_006230412.2 | XP_006230474.1 | 5 | 477 | |||
| XM_006230413.2 | XP_006230475.1 | 5 | 476 | |||
| XM_006230415.2 | XP_006230477.1 | 5 | 476 | |||
| XM_008759907.1 | XP_008758129.1 | 5 | 551 | |||
| XM_008759908.1 | XP_008758130.1 | 5 | 872 | |||
| XM_008759909.1 | XP_008758131.1 | 5 | 475 | |||
| XM_008759910.1 | XP_008758132.1 | 2 | 115 | |||
| GenBank mRNA | BC128752.1 | - | 5 | 428 | 7100072 | |

Plekha1
Genomic Map