siRNA Details
| Gene Symbol: Pstk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271229.1 | NP_001258158.1 | 2, 3 | 545 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pstk| Gene Symbol: Pstk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271229.1 | NP_001258158.1 | 2, 3 | 545 | ||