siRNA Details
| Gene Symbol: Ifitm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136124.1 | NP_001129596.1 | 2 | 411 | ||
| GenBank mRNA | AF164039.1 | - | 1 | 902 | ||
| BC168774.1 | - | 2 | 411 | 6921753 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifitm3| Gene Symbol: Ifitm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136124.1 | NP_001129596.1 | 2 | 411 | ||
| GenBank mRNA | AF164039.1 | - | 1 | 902 | ||
| BC168774.1 | - | 2 | 411 | 6921753 | ||