siRNA Details
| Gene Symbol: Cend1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014163.1 | NP_001014185.1 | 2 | 537 | ||
| GenBank mRNA | AB290916.1 | - | 2 | 455 | ||
| BC089963.1 | - | 2 | 537 | 7314412 | ||
| Gene Symbol: LOC100911402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749007.3 | XP_003749055.1 | 784 | |||

Cend1
Genomic Map