siRNA Details
| Gene Symbol: Tollip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109668.1 | NP_001103138.1 | 6 | 1963 | ||
| XM_006230607.2 | XP_006230669.1 | 7 | 2044 | |||
| XM_008760003.1 | XP_008758225.1 | 6 | 1908 | |||
| GenBank mRNA | BC133067.1 | - | 6 | 1849 | 7193007 | |

Tollip
Genomic Map