siRNA Details
| Gene Symbol: Dusp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108510.1 | NP_001101980.1 | 4 | 852 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp8| Gene Symbol: Dusp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108510.1 | NP_001101980.1 | 4 | 852 | ||