siRNA Details
| Gene Symbol: Tssc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013194.1 | NP_001013212.1 | 2 | 928 | ||
| XM_006230820.1 | XP_006230882.1 | 3 | 2041 | |||
| XM_006230821.1 | XP_006230883.1 | 2 | 2260 | |||
| XM_006230822.1 | XP_006230884.1 | 3 | 2011 | |||
| XM_008760148.1 | XP_008758370.1 | 2 | 2290 | |||
| XM_008760149.1 | XP_008758371.1 | 2 | 1946 | |||
| GenBank mRNA | BC083775.1 | - | 2 | 928 | 7193466 | |

Tssc4
Genomic Map