siRNA Details
| Gene Symbol: Pitpnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008369.1 | NP_001008370.1 | 10 | 1666 | ||
| GenBank mRNA | BC085945.1 | - | 10 | 1666 | 7128360 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pitpnm1| Gene Symbol: Pitpnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008369.1 | NP_001008370.1 | 10 | 1666 | ||
| GenBank mRNA | BC085945.1 | - | 10 | 1666 | 7128360 | |