siRNA Details
| Gene Symbol: Tmem134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001078647.1 | NP_001072115.1 | 1 | 178 | ||
| NM_001078648.1 | NP_001072116.1 | 1 | 178 | |||
| XM_006230835.2 | XP_006230897.1 | 1 | 231 | |||
| XM_006230837.2 | XP_006230899.1 | 1 | 235 | |||
| GenBank mRNA | BP495609.1 | - | 1 | 178 | ||
| FQ227694.1 | - | 1 | 173 | |||

Tmem134
Genomic Map