siRNA Details
| Gene Symbol: SUCLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003850.2 | NP_003841.1 | 9 | 1211 | ||
| XM_017020806.1 | XP_016876295.1 | 9 | 1431 | |||
| XM_017020807.1 | XP_016876296.1 | 11 | 1446 | |||
| XM_017020809.1 | XP_016876298.1 | 10 | 1295 | |||
| GenBank mRNA | AB035863.1 | - | 9 | 1211 | ||
| AF058953.1 | - | 8 | 1043 | |||
| AF087890.1 | - | 9 | 1219 | |||
| AK001458.1 | - | 9 | 1109 | |||
| AK001847.1 | - | 9 | 1198 | |||
| AK093506.1 | - | 3 | 1525 | |||
| AK225452.1 | - | 9 | 1198 | |||
| AK299437.1 | - | 9 | 1150 | |||
| AK302311.1 | - | 10 | 1270 | |||
| AK315513.1 | - | 9 | 1173 | |||
| AK316041.1 | - | 10 | 1125 | |||
| AK316271.1 | - | 10 | 1526 | |||
| AL080096.2 | - | 4 | 400 | |||
| BC027587.2 | - | 9 | 1163 | 4829291 | ||
| BX640602.1 | - | 9 | 1182 | |||
| KU178442.1 | - | 9 | 1154 | |||
| KU178443.1 | - | 6 | 710 | |||

SUCLA2
Genomic Map