siRNA Details
| Gene Symbol: Rps6kb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010962.2 | NP_001010962.2 | 5 | 439 | ||
| GenBank mRNA | BC089102.1 | - | 5 | 418 | 7305323 | |
| DY315655.1 | - | 491 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rps6kb2| Gene Symbol: Rps6kb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010962.2 | NP_001010962.2 | 5 | 439 | ||
| GenBank mRNA | BC089102.1 | - | 5 | 418 | 7305323 | |
| DY315655.1 | - | 491 | ||||