siRNA Details
| Gene Symbol: Pold4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013195.1 | NP_001013213.1 | 4 | 534 | ||
| GenBank mRNA | BC062014.1 | - | 4 | 534 | 5621348 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pold4| Gene Symbol: Pold4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013195.1 | NP_001013213.1 | 4 | 534 | ||
| GenBank mRNA | BC062014.1 | - | 4 | 534 | 5621348 | |