siRNA Details
| Gene Symbol: Ankrd13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108514.1 | NP_001101984.1 | 8 | 1014 | ||
| XM_006230842.2 | XP_006230904.1 | 8 | 1041 | |||
| GenBank mRNA | FQ213072.1 | - | 3 | 283 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd13d| Gene Symbol: Ankrd13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108514.1 | NP_001101984.1 | 8 | 1014 | ||
| XM_006230842.2 | XP_006230904.1 | 8 | 1041 | |||
| GenBank mRNA | FQ213072.1 | - | 3 | 283 | ||