siRNA Details
| Gene Symbol: Ctsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034110.1 | NP_001029282.1 | 5 | 537 | ||
| GenBank mRNA | BC099780.1 | - | 5 | 537 | 7933625 | |
| EU253481.1 | - | 5 | 567 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsf| Gene Symbol: Ctsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034110.1 | NP_001029282.1 | 5 | 537 | ||
| GenBank mRNA | BC099780.1 | - | 5 | 537 | 7933625 | |
| EU253481.1 | - | 5 | 567 | |||