siRNA Details
| Gene Symbol: Scgb2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203333.2 | NP_976078.1 | 2 | 241 | ||
| XM_008760232.1 | XP_008758454.1 | 2 | 540 | |||
| GenBank mRNA | BC062401.1 | - | 2 | 201 | 5621455 | |
| CB791758.1 | - | 2 | 253 | |||
| J00772.1 | - | 2 | 192 | |||
| V01263.1 | - | 2 | 216 | |||

Scgb2a2
Genomic Map