siRNA Details
| Gene Symbol: Sdhaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008371.1 | NP_001008372.1 | 2 | 187 | ||
| XM_006231042.1 | XP_006231104.1 | 2 | 284 | |||
| XM_006231043.2 | XP_006231105.1 | 2 | 242 | |||
| GenBank mRNA | BC076396.1 | - | 2 | 140 | ||
| BC086542.1 | - | 2 | 187 | 7122954 | ||

Sdhaf2
Genomic Map