siRNA Details
| Gene Symbol: Tmem216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271039.1 | NP_001257968.1 | 3 | 206 | ||
| XM_006231045.2 | XP_006231107.1 | 3 | 1031 | |||
| GenBank mRNA | BC168207.1 | - | 2 | 175 | 7131074 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem216| Gene Symbol: Tmem216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271039.1 | NP_001257968.1 | 3 | 206 | ||
| XM_006231045.2 | XP_006231107.1 | 3 | 1031 | |||
| GenBank mRNA | BC168207.1 | - | 2 | 175 | 7131074 | |