siRNA Details
| Gene Symbol: Cyb561a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014164.1 | NP_001014186.1 | 6 | 928 | ||
| XM_006231050.2 | XP_006231112.1 | 6 | 1396 | |||
| XM_006231051.2 | XP_006231113.1 | 6 | 1361 | |||
| XM_006231052.2 | XP_006231114.1 | 6 | 1563 | |||
| XM_006231053.2 | XP_006231115.1 | 6 | 947 | |||
| XM_006231054.2 | XP_006231116.1 | 6 | 907 | |||
| XM_006231055.2 | XP_006231117.1 | 5 | 1110 | |||
| XM_008760233.1 | XP_008758455.1 | 6 | 1283 | |||
| XM_008760234.1 | XP_008758456.1 | 6 | 891 | |||
| GenBank mRNA | BC085835.1 | - | 6 | 928 | 7134938 | |

Cyb561a3
Genomic Map