siRNA Details
Gene Symbol: Tmem252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007738.1 | NP_001007739.1 | 2 | 497 | ||
GenBank mRNA | BC079347.1 | - | 2 | 497 | 7107289 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007738.1 | NP_001007739.1 | 2 | 497 | ||
GenBank mRNA | BC079347.1 | - | 2 | 497 | 7107289 |