siRNA Details
| Gene Symbol: Il33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014166.1 | NP_001014188.1 | 3 | 226 | ||
| XM_008760344.1 | XP_008758566.1 | 3 | 249 | |||
| GenBank mRNA | BC081993.1 | - | 3 | 226 | 7130307 | |
| FQ212643.1 | - | 3 | 254 | |||

Il33
Genomic Map