siRNA Details
| Gene Symbol: Opalin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017386.1 | NP_001017386.1 | 5, 6 | 281 | ||
| XM_006231401.2 | XP_006231463.1 | 3, 4 | 1122 | |||
| GenBank mRNA | BC092200.1 | - | 5, 6 | 281 | 7319643 | |

Opalin
Genomic Map