siRNA Details
| Gene Symbol: Opalin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017386.1 | NP_001017386.1 | 6 | 340 | ||
| XM_006231401.2 | XP_006231463.1 | 4 | 1181 | |||
| GenBank mRNA | BC092200.1 | - | 6 | 340 | 7319643 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Opalin| Gene Symbol: Opalin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017386.1 | NP_001017386.1 | 6 | 340 | ||
| XM_006231401.2 | XP_006231463.1 | 4 | 1181 | |||
| GenBank mRNA | BC092200.1 | - | 6 | 340 | 7319643 | |