siRNA Details
Gene Symbol: Cutc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231526.1 | XP_006231588.1 | 5, 6 | 569 | ||
XM_006231527.1 | XP_006231589.1 | 4, 5 | 533 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cutc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231526.1 | XP_006231588.1 | 5, 6 | 569 | ||
XM_006231527.1 | XP_006231589.1 | 4, 5 | 533 |