siRNA Details
Gene Symbol: Btrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007148.2 | NP_001007149.2 | 9 | 1073 | ||
XM_006231529.2 | XP_006231591.1 | 9 | 1656 | |||
XM_006231530.2 | XP_006231592.1 | 8 | 1067 | |||
XM_006231531.2 | XP_006231593.1 | 8 | 1033 | |||
XM_006231535.2 | XP_006231597.1 | 8 | 918 | |||
XM_006231536.2 | XP_006231598.1 | 8 | 1000 | |||
XM_006231537.2 | XP_006231599.1 | 8 | 1547 | |||
XM_006231538.2 | XP_006231600.1 | 7 | 957 | |||
XM_006231539.2 | XP_006231601.1 | 7 | 1703 | |||
XM_006231541.2 | XP_006231603.1 | 7 | 1417 | |||
XM_006231543.2 | XP_006231605.1 | 6 | 1625 | |||
GenBank mRNA | BC085125.1 | - | 8 | 894 | 7131883 |
