siRNA Details
| Gene Symbol: Btrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007148.2 | NP_001007149.2 | 14, 15 | 1799 | ||
| XM_006231529.2 | XP_006231591.1 | 14, 15 | 2382 | |||
| XM_006231530.2 | XP_006231592.1 | 13, 14 | 1793 | |||
| XM_006231531.2 | XP_006231593.1 | 13, 14 | 1759 | |||
| XM_006231535.2 | XP_006231597.1 | 13, 14 | 1644 | |||
| XM_006231536.2 | XP_006231598.1 | 13, 14 | 1726 | |||
| XM_006231537.2 | XP_006231599.1 | 13, 14 | 2273 | |||
| XM_006231538.2 | XP_006231600.1 | 12, 13 | 1683 | |||
| XM_006231539.2 | XP_006231601.1 | 12, 13 | 2429 | |||
| XM_006231541.2 | XP_006231603.1 | 12, 13 | 2143 | |||
| XM_006231543.2 | XP_006231605.1 | 11, 12 | 2351 | |||
| GenBank mRNA | BC085125.1 | - | 13, 14 | 1620 | 7131883 | |

Btrc
Genomic Map