siRNA Details
Gene Symbol: Btrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007148.2 | NP_001007149.2 | 15 | 1844 | ||
XM_006231529.2 | XP_006231591.1 | 15 | 2427 | |||
XM_006231530.2 | XP_006231592.1 | 14 | 1838 | |||
XM_006231531.2 | XP_006231593.1 | 14 | 1804 | |||
XM_006231535.2 | XP_006231597.1 | 14 | 1689 | |||
XM_006231536.2 | XP_006231598.1 | 14 | 1771 | |||
XM_006231537.2 | XP_006231599.1 | 14 | 2318 | |||
XM_006231538.2 | XP_006231600.1 | 13 | 1728 | |||
XM_006231539.2 | XP_006231601.1 | 13 | 2474 | |||
XM_006231541.2 | XP_006231603.1 | 13 | 2188 | |||
XM_006231543.2 | XP_006231605.1 | 12 | 2396 | |||
GenBank mRNA | BC085125.1 | - | 14 | 1665 | 7131883 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001286466, NM_001286465, NM_001037758, NM_009771, AK032221, and others.
Gene Ontology Categories:
