siRNA Details
| Gene Symbol: Zdhhc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037652.1 | NP_001032741.1 | 1127 | |||
| GenBank mRNA | AY886524.1 | - | 5, 6 | 1008 | ||
| BC107928.1 | - | 1127 | 7130799 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zdhhc6| Gene Symbol: Zdhhc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037652.1 | NP_001032741.1 | 1127 | |||
| GenBank mRNA | AY886524.1 | - | 5, 6 | 1008 | ||
| BC107928.1 | - | 1127 | 7130799 | |||