siRNA Details
| Gene Symbol: Ccdc172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014169.2 | NP_001014191.1 | 6 | 590 | ||
| XM_006231674.2 | XP_006231736.1 | 6 | 606 | |||
| XM_008760539.1 | XP_008758761.1 | 5 | 879 | |||
| XM_008760540.1 | XP_008758762.1 | 4 | 762 | |||
| XM_008760541.1 | XP_008758763.1 | 5 | 879 | |||
| XM_008760542.1 | XP_008758764.1 | 5 | 879 | |||
| GenBank mRNA | BC079324.1 | - | 5 | 807 | 7135457 | |

Ccdc172
Genomic Map