siRNA Details
| Gene Symbol: Sfxn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108527.1 | NP_001101997.1 | 14 | 997 | ||
| XM_006231684.2 | XP_006231746.1 | 13 | 3058 | |||
| XM_006231685.2 | XP_006231747.1 | 13 | 1894 | |||
| XM_008760590.1 | XP_008758812.1 | 12 | 2792 | |||

Sfxn4
Genomic Map