siRNA Details
| Gene Symbol: ADSS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126.3 | NP_001117.2 | 12 | 1606 | ||
| XM_011544108.2 | XP_011542410.1 | 12 | 2278 | |||
| GenBank mRNA | AK025514.1 | - | 12 | 1353 | ||
| AK303886.1 | - | 13 | 1318 | |||
| BC012356.1 | - | 12 | 1391 | 4635347 | ||

ADSS
Genomic Map