siRNA Details
| Gene Symbol: Atat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212498.1 | NP_997663.1 | 6 | 499 | ||
| XM_006255948.2 | XP_006256010.1 | 6 | 534 | |||
| XM_006255950.2 | XP_006256012.1 | 6 | 534 | |||
| XM_006255951.2 | XP_006256013.1 | 6 | 534 | |||
| XM_008772731.1 | XP_008770953.1 | 6 | 534 | |||

Atat1
Genomic Map