siRNA Details
| Gene Symbol: Ppp1r18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126287.2 | NP_001119759.2 | 2 | 762 | ||
| XM_006255952.2 | XP_006256014.1 | 1 | 2486 | |||
| XM_006255953.2 | XP_006256015.1 | 2 | 1554 | |||
| GenBank mRNA | BC161960.1 | - | 2 | 762 | 9027646 | |

Ppp1r18
Genomic Map