siRNA Details
| Gene Symbol: Rxrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206849.3 | NP_996731.2 | 4 | 725 | ||
| XM_006255960.2 | XP_006256022.1 | 4 | 1235 | |||
| XM_006255961.2 | XP_006256023.1 | 4 | 728 | |||
| GenBank mRNA | BC099776.1 | - | 4 | 627 | ||
| M81766.1 | - | 4 | 554 | |||

Rxrb
Genomic Map