siRNA Details
| Gene Symbol: Lemd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039032.1 | NP_001034121.1 | 1 | 307 | ||
| GenBank mRNA | BC098775.1 | - | 1 | 307 | 7373953 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lemd2| Gene Symbol: Lemd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039032.1 | NP_001034121.1 | 1 | 307 | ||
| GenBank mRNA | BC098775.1 | - | 1 | 307 | 7373953 | |