siRNA Details
| Gene Symbol: Zfp523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134755.1 | NP_001128227.1 | 12, 13 | 1609 | ||
| XM_008772820.1 | XP_008771042.1 | 11, 12 | 1592 | |||
| XM_008772821.1 | XP_008771043.1 | 11, 12 | 1586 | |||
| GenBank mRNA | BC168249.1 | - | 12, 13 | 1609 | 5598849 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_172617, AK028222, BC058346, BC060611, AK186832, and others.

Zfp523
Genomic Map