siRNA Details
Gene Symbol: Zfp523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134755.1 | NP_001128227.1 | 6, 7 | 655 | ||
XM_008772820.1 | XP_008771042.1 | 5, 6 | 638 | |||
XM_008772821.1 | XP_008771043.1 | 5, 6 | 632 | |||
GenBank mRNA | BC168249.1 | - | 6, 7 | 655 | 5598849 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
