siRNA Details
| Gene Symbol: Fkbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012174.1 | NP_001012174.1 | 4 | 492 | ||
| XM_006256222.2 | XP_006256284.1 | 4 | 1241 | |||
| XM_006256224.2 | XP_006256286.1 | 5 | 1308 | |||
| GenBank mRNA | BC085868.1 | - | 4 | 492 | 7191081 | |

Fkbp5
Genomic Map