siRNA Details
| Gene Symbol: CDKL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282236.1 | NP_001269165.1 | 7 | 1355 | ||
| NM_004196.4 | NP_004187.2 | 6 | 763 | |||
| XM_005268157.2 | XP_005268214.1 | 7 | 1355 | |||
| XM_005268159.2 | XP_005268216.1 | 7 | 1355 | |||
| XM_005268160.4 | XP_005268217.1 | 4 | 2451 | |||
| XM_011537276.1 | XP_011535578.1 | 7 | 765 | |||
| XM_017021729.1 | XP_016877218.1 | 7 | 1247 | |||
| XM_017021730.1 | XP_016877219.1 | 7 | 1247 | |||
| XM_017021731.1 | XP_016877220.1 | 7 | 1256 | |||
| XM_017021732.1 | XP_016877221.1 | 7 | 1247 | |||
| GenBank mRNA | AF390028.1 | - | 19 | 2723 | ||
| AK308732.1 | - | 7 | 1350 | |||
| BC104977.1 | - | 6 | 738 | 8143980 | ||
| X66358.1 | - | 6 | 923 | |||

CDKL1
Genomic Map