siRNA Details
| Gene Symbol: Ube2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108530.1 | NP_001102000.1 | 5, 6 | 404 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2d1| Gene Symbol: Ube2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108530.1 | NP_001102000.1 | 5, 6 | 404 | ||