siRNA Details
| Gene Symbol: Ank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033984.1 | NP_001029156.1 | 43 | 7548 | ||
| NM_031805.1 | NP_113993.1 | 39 | 5782 | |||
| GenBank mRNA | AF069525.1 | - | 39 | 5052 | ||
| AF102552.1 | - | 43 | 7548 | |||
| AJ428573.1 | - | 18 | 2377 | |||
| AJ812019.1 | - | 18 | 2737 | |||
| AJ812020.1 | - | 18 | 2737 | |||
| AJ812021.1 | - | 18 | 3325 | |||
| AJ812022.1 | - | 18 | 3325 | |||
| AJ812023.1 | - | 39 | 5186 | |||
| AJ812024.1 | - | 39 | 5774 | |||
| BC101885.1 | - | 18 | 2418 | |||

Ank3
Genomic Map