siRNA Details
| Gene Symbol: RGD1306739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134576.1 | NP_001128048.1 | 7 | 871 | ||
| XM_006256355.2 | XP_006256417.1 | 7 | 902 | |||
| XM_006256356.2 | XP_006256418.1 | 7 | 877 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1306739| Gene Symbol: RGD1306739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134576.1 | NP_001128048.1 | 7 | 871 | ||
| XM_006256355.2 | XP_006256417.1 | 7 | 902 | |||
| XM_006256356.2 | XP_006256418.1 | 7 | 877 | |||