siRNA Details
| Gene Symbol: Sar1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007739.1 | NP_001007740.1 | 5 | 475 | ||
| XM_006256450.2 | XP_006256512.1 | 6 | 469 | |||
| XM_006256451.2 | XP_006256513.1 | 5 | 493 | |||
| GenBank mRNA | BC079228.1 | - | 5 | 475 | 7132455 | |
| FQ232554.1 | - | 5 | 429 | |||

Sar1a
Genomic Map