siRNA Details
| Gene Symbol: Eif4ebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033069.1 | NP_001028241.1 | 1, 2 | 266 | ||
| XM_008772891.1 | XP_008771113.1 | 1, 2 | 204 | |||
| GenBank mRNA | BC100638.1 | - | 1, 2 | 266 | 5600115 | |

Eif4ebp2
Genomic Map