siRNA Details
| Gene Symbol: Vps26a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007740.1 | NP_001007741.1 | 5 | 505 | ||
| XM_006256460.2 | XP_006256522.1 | 5 | 628 | |||
| XM_008772892.1 | XP_008771114.1 | 5 | 633 | |||
| GenBank mRNA | BC079150.1 | - | 5 | 505 | 7128580 | |
| FQ211877.1 | - | 5 | 518 | |||

Vps26a
Genomic Map