siRNA Details
| Gene Symbol: Ddx50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013198.2 | NP_001013216.2 | 15 | 2245 | ||
| GenBank mRNA | BC091427.1 | - | 9 | 1131 | 7366491 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx50| Gene Symbol: Ddx50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013198.2 | NP_001013216.2 | 15 | 2245 | ||
| GenBank mRNA | BC091427.1 | - | 9 | 1131 | 7366491 | |